Gene Transcripts

Gene Symbol
Smg8
Entrez Gene ID
74133
Gene Name
smg-8 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Aliases
1200011M11Rik
Chromosome Location
Chr. 11: 87077732 - 87086777 on Build GRCm38
UniGene ID
Mm.23257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024262.1 NP_077224.1 1 888
GenBank mRNA AK004714.1 - 1 881
AK031106.1 - 1 862
BC020005.1 - 1 888 3981459

View Full Product Details