Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027902.2 | NP_082178.2 | 7 | 979 | |
GenBank mRNA | AK004939.2 | - | 7 | 826 | |
AY240929.1 | - | 7 | 956 | ||
BC029645.1 | - | 7 | 956 | 4195486 | |
BC057674.1 | - | 7 | 921 | 4162007 | |
EU190436.1 | - | 6 | 770 | ||
EU190437.1 | - | 6 | 770 |