Gene Transcripts

Gene Symbol
Endod1
Entrez Gene ID
71946
Gene Name
endonuclease domain containing 1
Gene Aliases
2210414F18Rik, 2310067E08Rik, C85344
Chromosome Location
Chr. 9: 14353990 - 14381242 on Build GRCm38
UniGene ID
Mm.41423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028013.3 NP_082289.2 2 709
GenBank mRNA AK078056.1 - 2 460
AK079741.1 - 2 448
AK086102.1 - 2 466
AK142446.1 - 3 387
AK173036.1 - 2 709
BC096490.1 - 2 440 6439018

View Full Product Details