Gene Transcripts

Gene Symbol
Endod1
Entrez Gene ID
71946
Gene Name
endonuclease domain containing 1
Gene Aliases
2210414F18Rik, 2310067E08Rik, C85344
Chromosome Location
Chr. 9: 14353990 - 14381242 on Build GRCm38
UniGene ID
Mm.41423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028013.3 NP_082289.2 2 1797
GenBank mRNA AK010084.1 - 1 379
AK078056.1 - 2 1548
AK079741.1 - 2 1536
AK086102.1 - 2 1555
AK142446.1 - 3 1475
AK173036.1 - 2 1797
AK192560.1 - 1 42
BC019813.1 - 2 1563 4017632
BC096490.1 - 2 1528 6439018

View Full Product Details