Gene Transcripts

Gene Symbol
Ist1
Entrez Gene ID
71955
Gene Name
increased sodium tolerance 1 homolog (yeast)
Gene Aliases
2400003C14Rik, AW536298, mKIAA0174
Chromosome Location
Chr. 8: 109671321 - 109693294 on Build GRCm38
UniGene ID
Mm.290036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028018.2 NP_082294.1 3, 4 404
GenBank mRNA AK010262.1 - 3, 4 374
AK050468.1 - 3, 4 371
AK088192.1 - 3, 4 359
AK122215.1 - 1, 2 178
AK152830.1 - 3, 4 371
AK161899.1 - 3, 4 360
AK207302.1 - 1, 2 98
AK218302.1 - 1, 2 36
BC017605.1 - 3, 4 370 4507347
BC039052.1 - 3, 4 347 2631157

View Full Product Details