Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163032.1 | NP_001156504.1 | 5 | 2063 | |
NM_028052.4 | NP_082328.3 | 6 | 1957 | ||
GenBank mRNA | AK005132.1 | - | 6 | 1878 | |
AK032442.1 | - | 5 | 1965 | ||
AK049661.1 | - | 5 | 1909 | ||
AK078223.1 | - | 6 | 1958 | ||
BC026512.1 | - | 6 | 1904 | 4527827 |