Gene Transcripts
- Gene Symbol
- NPEPPS
- Entrez Gene ID
- 9520
- Gene Name
- aminopeptidase puromycin sensitive
- Gene Aliases
- AAP-S, MP100, PSA
- Chromosome Location
- Chr. 17: 47522950 - 47623276 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
- Gene Symbol
- LOC101929950
- Entrez Gene ID
- 101929950
- Gene Name
- puromycin-sensitive aminopeptidase-like protein
- Gene Aliases
- Chromosome Location
- Chr. 17: 38078159 - 38118364 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC440434
- Entrez Gene ID
- 440434
- Gene Name
- aminopeptidase puromycin sensitive pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 38195740 - 38257249 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036750.2 |
|
2 |
369 |
|
View Full Product Details