Gene Transcripts

Gene Symbol
Atp13a2
Entrez Gene ID
74772
Gene Name
ATPase type 13A2
Gene Aliases
1110012E06Rik, AA589443
Chromosome Location
Chr. 4: 140986865 - 141007701 on Build GRCm38
UniGene ID
Mm.205625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164366.1 NP_001157838.1 5, 6 667
NM_029097.2 NP_083373.2 5, 6 667
GenBank mRNA AK156104.1 - 5, 6 647
AK171554.1 - 5 920
AK219794.1 - 1, 2 89
BC023746.1 - 5, 6 642 5353766
BC025028.1 - 5, 6 562
BC042661.1 - 5, 6 612 4192094

View Full Product Details