Gene Transcripts
- Gene Symbol
- Chmp4b
- Entrez Gene ID
- 75608
- Gene Name
- charged multivesicular body protein 4B
- Gene Aliases
- 2010012F05Rik, C76846, Snf7-2
- Chromosome Location
- Chr. 2: 154657026 - 154694783 on Build GRCm38
- UniGene ID
- Mm.262480
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC011429.1 |
- |
1 |
312 |
|
BC059279.1 |
- |
1 |
287 |
4019739 |
View Full Product Details