Gene Transcripts

Gene Symbol
Klf17
Entrez Gene ID
75753
Gene Name
Kruppel-like factor 17
Gene Aliases
7420700M05Rik, AA420409, AU043468, C85123, D4Ertd561e, Gzf, Zfp393
Chromosome Location
Chr. 4: 117756659 - 117765666 on Build GRCm38
UniGene ID
Mm.3848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029416.2 NP_083692.2 2 724
GenBank mRNA AF508984.1 - 2 690
AK018361.1 - 2 690
AK139827.1 - 2 691
BC064748.1 - 2 668 30476790

View Full Product Details