Gene Transcripts

Gene Symbol
Pcgf5
Entrez Gene ID
76073
Gene Name
polycomb group ring finger 5
Gene Aliases
0610009F02Rik, 1110054A01Rik, 5830406C17Rik, 5830443C21Rik, 9530023M17Rik, AI324127
Chromosome Location
Chr. 19: 36348084 - 36460970 on Build GRCm38
UniGene ID
Mm.259180
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029508.3 NP_083784.1 8, 9 925
GenBank mRNA AK002387.1 - 8, 9 949
AK017984.1 - 8, 9 926
BC139258.1 - 8, 9 886 8862280
BC139259.1 - 8, 9 886 8862281

View Full Product Details