Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133733.4 | NP_598494.2 | 3 | 579 | |
GenBank mRNA | AB040490.1 | - | 3 | 496 | |
AK028983.1 | - | 3 | 549 | ||
AK033723.1 | - | 3 | 581 | ||
AK080481.1 | - | 3 | 534 | ||
AY259213.1 | - | 4 | 208 | ||
AY326421.1 | - | 3 | 214 | ||
BC026447.1 | - | 3 | 405 | 4459717 |