Gene Transcripts

Gene Symbol
Ttc27
Entrez Gene ID
74196
Gene Name
tetratricopeptide repeat domain 27
Gene Aliases
2610511O17Rik
Chromosome Location
Chr. 17: 74717736 - 74863570 on Build GRCm38
UniGene ID
Mm.209503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152817.4 NP_690030.3 13 1680
GenBank mRNA AK012131.1 - 13 1680
AK030942.1 - 13 1681
AK049324.1 - 13 1681
AK168138.1 - 13 1671
BC021912.1 - 12 1415 4503335

View Full Product Details