Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152818.2 | NP_690031.2 | 14 | 3825 | |
GenBank mRNA | AK044598.1 | - | 11 | 2616 | |
BC031794.1 | - | 12 | 2788 | 4501119 | |
BC034567.1 | - | 4 | 1665 | ||
BC058356.1 | - | 13 | 2968 | 5696357 |