Gene Transcripts

Gene Symbol
Tonsl
Entrez Gene ID
72749
Gene Name
tonsoku-like, DNA repair protein
Gene Aliases
2810439M11Rik, Nfkbil2
Chromosome Location
Chr. 15: 76626237 - 76639929 on Build GRCm38
UniGene ID
Mm.358749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183091.3 NP_898914.3 8 984
GenBank mRNA AK139173.1 - 8 985
BC066068.1 - 8 966 6851399

View Full Product Details