Gene Transcripts
- Gene Symbol
- Col4a3bp
- Entrez Gene ID
- 68018
- Gene Name
- collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
- Gene Aliases
- 2810404O15Rik, 9230101K08Rik, AU016711, CERT, GPBP
- Chromosome Location
- Chr. 13: 96542735 - 96640167 on Build GRCm38
- UniGene ID
- Mm.24125
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164222.1 |
NP_001157694.1
|
2, 3 |
667 |
|
NM_023420.2 |
NP_075909.1
|
2, 3 |
667 |
|
GenBank mRNA |
AF232932.1 |
- |
2, 3 |
681 |
|
AF232934.1 |
- |
2, 3 |
681 |
|
AK012989.1 |
- |
2, 3 |
648 |
|
AK030305.1 |
- |
3, 4 |
575 |
|
AK038061.1 |
- |
2, 3 |
657 |
|
AK038803.1 |
- |
2, 3 |
648 |
|
AK047100.1 |
- |
2, 3 |
664 |
|
AK080958.1 |
- |
2, 3 |
665 |
|
AK163624.1 |
- |
2, 3 |
657 |
|
BC016197.1 |
- |
2, 3 |
630 |
4502118 |
View Full Product Details