Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023557.3 | NP_076046.1 | 5 | 273 | |
GenBank mRNA | AK145621.1 | - | 5 | 265 | |
AK164684.1 | - | 4 | 917 | ||
AK166620.1 | - | 2 | 175 | ||
BC010808.1 | - | 5 | 271 | 4240658 | |
BC052652.1 | - | 5 | 271 | 30034507 |