Gene Transcripts

Gene Symbol
Rab3c
Entrez Gene ID
67295
Gene Name
RAB3C, member RAS oncogene family
Gene Aliases
2700062I01Rik, 3110015B08Rik, 3110037E15Rik, AI850886
Chromosome Location
Chr. 13: 110054187 - 110281041 on Build GRCm38
UniGene ID
Mm.151600
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023852.5 NP_076341.1 5 1028
GenBank mRNA AK014132.1 - 5 950
AK079574.1 - 5 925
AK148904.1 - 5 949
AK161006.1 - 5 926
AK164316.1 - 5 973
AY026947.1 - 5 909
BC114335.2 - 5 901 40045744

View Full Product Details