Gene Transcripts

Gene Symbol
1700028P14Rik
Entrez Gene ID
67483
Gene Name
RIKEN cDNA 1700028P14 gene
Gene Aliases
AV209940
Chromosome Location
Chr. 19: 23558760 - 23652812 on Build GRCm38
UniGene ID
Mm.443287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026188.2 NP_080464.1 2 310
GenBank mRNA AK006473.1 - 2 313
AK014972.1 - 2 312
BC049563.1 - 2 295 6771302

View Full Product Details