Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
67865
Gene Name
regulator of G-protein signalling 10
Gene Aliases
2310010N19Rik
Chromosome Location
Chr. 7: 128373621 - 128419061 on Build GRCm38
UniGene ID
Mm.18635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026418.2 NP_080694.1 5 845
GenBank mRNA AK009283.1 - 5 846
AK009686.1 - 5 847
AK021202.1 - 5 849
AK088009.1 - 5 844
EF410104.1 - 642

View Full Product Details