Gene Transcripts
- Gene Symbol
- GCGR
- Entrez Gene ID
- 2642
- Gene Name
- glucagon receptor
- Gene Aliases
- GGR, GL-R
- Chromosome Location
- Chr. 17: 81804078 - 81814013 on Build GRCh38
- UniGene ID
- Hs.208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000160.4 |
NP_000151.1
|
5, 6 |
681 |
|
XM_006722277.1 |
XP_006722340.1
|
5, 6 |
710 |
|
XM_011523539.1 |
XP_011521841.1
|
5, 6 |
808 |
|
XM_017024446.1 |
XP_016879935.1
|
5, 6 |
729 |
|
XM_017024447.1 |
XP_016879936.1
|
4, 5 |
518 |
|
GenBank mRNA |
AK314247.1 |
- |
5, 6 |
664 |
|
BC104854.1 |
- |
4, 5 |
431 |
8143857 |
BC112041.1 |
- |
4, 5 |
430 |
8327509 |
L20316.1 |
- |
5, 6 |
663 |
|
U03469.1 |
- |
4, 5 |
447 |
|
View Full Product Details