Gene Transcripts

Gene Symbol
Kptn
Entrez Gene ID
70394
Gene Name
kaptin
Gene Aliases
2310042D10Rik, 2E4, C030013F01Rik
Chromosome Location
Chr. 7: 16119876 - 16127516 on Build GRCm38
UniGene ID
Mm.41101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133727.2 NP_598488.2 10, 11 1090
GenBank mRNA AK009746.1 - 10, 11 1071
BC018316.1 - 10, 11 1058 4160465

View Full Product Details