Gene Transcripts

Gene Symbol
Tra2a
Entrez Gene ID
101214
Gene Name
transformer 2 alpha homolog (Drosophila)
Gene Aliases
1500010G04Rik, AL022798, G430041M01Rik, mAWMS1
Chromosome Location
Chr. 6: 49243921 - 49264248 on Build GRCm38
UniGene ID
Mm.196598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB052758.1 - 1 112
AK005187.1 - 1 823
AK076230.1 - 1 807
AK076606.1 - 1 791
AK148552.1 - 2 574

View Full Product Details