Gene Transcripts

Gene Symbol
Wipf2
Entrez Gene ID
68524
Gene Name
WAS/WASL interacting protein family, member 2
Gene Aliases
1110014J05Rik, 5730509C05Rik, AA407487, Gm1176, Wich, Wire
Chromosome Location
Chr. 11: 98863598 - 98905578 on Build GRCm38
UniGene ID
Mm.28661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197940.2 NP_922922.1 8 4288
GenBank mRNA AK044867.1 - 1 1432
AK046226.1 - 1 2608
BC057854.1 - 8 4077 6494304

View Full Product Details