Gene Transcripts

Gene Symbol
Trappc2l
Entrez Gene ID
59005
Gene Name
trafficking protein particle complex 2-like
Gene Aliases
1810017G16Rik, AB030200, Hspc176, Tca17
Chromosome Location
Chr. 8: 122611626 - 122615591 on Build GRCm38
UniGene ID
Mm.38746
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021502.2 NP_067477.1 4 349
GenBank mRNA AB030200.1 - 4 342
AK007526.1 - 4 319
BC028502.1 - 4 329 1366538
BC114968.1 - 317
BY091059.1 - 4 350

View Full Product Details