Gene Transcripts

Gene Symbol
Jph2
Entrez Gene ID
59091
Gene Name
junctophilin 2
Gene Aliases
1110002E14Rik, JP-2, Jp2
Chromosome Location
Chr. 2: 163336242 - 163398049 on Build GRCm38
UniGene ID
Mm.34459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205076.1 NP_001192005.1 3 758
NM_021566.2 NP_067541.1 2 838
GenBank mRNA AB024447.1 - 2 815
AK157145.1 - 3 758

View Full Product Details