Gene Transcripts
- Gene Symbol
- Sarnp
- Entrez Gene ID
- 66118
- Gene Name
- SAP domain containing ribonucleoprotein
- Gene Aliases
- 1110005A23Rik, Cip29
- Chromosome Location
- Chr. 10: 128821766 - 128877638 on Build GRCm38
- UniGene ID
- Mm.351579
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025364.2 |
NP_079640.1
|
4, 5 |
413 |
|
GenBank mRNA |
AK003453.1 |
- |
4, 5 |
265 |
|
AK018773.1 |
- |
4, 5 |
276 |
|
AK088266.1 |
- |
4, 5 |
262 |
|
AK146248.1 |
- |
4, 5 |
291 |
|
AK212934.1 |
- |
5, 6 |
297 |
|
BC027510.1 |
- |
4, 5 |
252 |
1382354 |
BC094393.1 |
- |
4, 5 |
249 |
|
BC157909.1 |
- |
4, 5 |
277 |
9088111 |
BC158023.1 |
- |
4, 5 |
277 |
9007329 |
BC172063.1 |
- |
4, 5 |
274 |
9054349 |
View Full Product Details