Gene Transcripts

Gene Symbol
Thoc7
Entrez Gene ID
66231
Gene Name
THO complex 7 homolog (Drosophila)
Gene Aliases
Nif3l1bp1
Chromosome Location
Chr. 14: 13949012 - 13962042 on Build GRCm38
UniGene ID
Mm.295875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285780.1 NP_001272709.1 5 527
NM_025435.3 NP_079711.1 4 449
GenBank mRNA AK005176.1 - 4 376
AK007440.1 - 4 355
AK010958.1 - 4 390
AK013319.1 - 4 372
AK143106.1 - 2 1597
AK147003.1 - 5 430
BC027113.1 - 3 1397
BC042209.1 - 4 376
BC054419.1 - 4 409 4010343
BC116919.1 - 4 414 40126238
BC116945.1 - 4 414 40126264
CK329337.1 - 4 450

View Full Product Details