Gene Transcripts

Gene Symbol
Lamtor1
Entrez Gene ID
66508
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Gene Aliases
2400001E08Rik, Pdro, p18
Chromosome Location
Chr. 7: 101905837 - 101911903 on Build GRCm38
UniGene ID
Mm.214841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025605.3 NP_079881.2 2 281
GenBank mRNA AK003509.1 - 2 173
AK009320.1 - 2 170
AK010247.1 - 2 214
AK013389.1 - 2 191
AK144476.1 - 2 180
AK185971.1 - 2 124
AK203171.1 - 2 123
BC020142.1 - 2 187 3709051
BC092062.1 - 2 173 4024506
BC096412.1 - 2 153 5255882
BC126973.1 - 2 188 40100355

View Full Product Details