Gene Transcripts

Gene Symbol
Rwdd1
Entrez Gene ID
66521
Gene Name
RWD domain containing 1
Gene Aliases
0710001K08Rik, 2610002D06Rik, 2700069A07Rik, IH1
Chromosome Location
Chr. 10: 33996555 - 34019616 on Build GRCm38
UniGene ID
Mm.28528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025614.3 NP_079890.1 2 287
GenBank mRNA AF503942.1 - 2 179
AK003262.2 - 2 241
AK005278.1 - 2 199
AK009510.1 - 2 156
AK011277.1 - 2 287
AK012504.1 - 2 258
BC062136.1 - 2 169 6508909

View Full Product Details