Gene Transcripts

Gene Symbol
Pomp
Entrez Gene ID
66537
Gene Name
proteasome maturation protein
Gene Aliases
2510048O06Rik, Ump1
Chromosome Location
Chr. 5: 147860628 - 147875778 on Build GRCm38
UniGene ID
Mm.332855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025624.2 NP_079900.1 3 207
GenBank mRNA AK003439.1 - 3 200
AK011115.1 - 3 208
AK012455.1 - 3 169
AK014296.1 - 3 171
AK044283.1 - 3 183
AK132041.1 - 184
AK167958.1 - 3 208
BC027016.1 - 3 198 5375452

View Full Product Details