Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025649.3 | NP_079925.2 | 3 | 1129 | |
GenBank mRNA | AK002377.1 | - | 3 | 1115 | |
AK004224.1 | - | 3 | 1083 | ||
AK007974.1 | - | 3 | 1081 | ||
AK011037.1 | - | 1087 | |||
AK156569.1 | - | 3 | 1132 | ||
BC005768.1 | - | 3 | 1087 | 2649017 |