Gene Transcripts
- Gene Symbol
- Golph3
- Entrez Gene ID
- 66629
- Gene Name
- golgi phosphoprotein 3
- Gene Aliases
- 4733401N08Rik, 5730410D03Rik, AW413496
- Chromosome Location
- Chr. 15: 12321197 - 12351601 on Build GRCm38
- UniGene ID
- Mm.157655
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025673.2 |
NP_079949.1
|
1, 2 |
463 |
|
GenBank mRNA |
AK005996.1 |
- |
1, 2 |
510 |
|
AK014644.1 |
- |
1, 2 |
511 |
|
AK015543.1 |
- |
1, 2 |
507 |
|
AK021327.1 |
- |
1, 2 |
509 |
|
AK028409.1 |
- |
1, 2 |
472 |
|
AK077446.1 |
- |
1, 2 |
528 |
|
AK133763.1 |
- |
1, 2 |
511 |
|
AK136194.1 |
- |
1, 2 |
88 |
|
AK167805.1 |
- |
1, 2 |
504 |
|
BC031445.1 |
- |
1, 2 |
324 |
4482612 |
BC039909.1 |
- |
1, 2 |
461 |
|
BC051035.1 |
- |
1, 2 |
463 |
2655316 |
View Full Product Details