Gene Transcripts

Gene Symbol
UTP14A
Entrez Gene ID
10813
Gene Name
UTP14A small subunit processome component
Gene Aliases
NYCO16, SDCCAG16, dJ537K23.3
Chromosome Location
Chr. X: 129906121 - 129929762 on Build GRCh38
UniGene ID
Hs.458598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166221.1 NP_001159693.1 10 914
NM_006649.3 NP_006640.2 11 1070
XM_017029237.1 XP_016884726.1 13 1222
XM_017029238.1 XP_016884727.1 11 1252
GenBank mRNA AK094408.1 - 2 66
AK291918.1 - 11 1037
AK298578.1 - 10 883
AK304563.1 - 10 935
BC001149.1 - 11 1006 3355662
BC009649.1 - 11 1008 3459192
BC014987.1 - 11 998 4901477

View Full Product Details