Gene Transcripts

Gene Symbol
Aspn
Entrez Gene ID
66695
Gene Name
asporin
Gene Aliases
4631401G09Rik, AA986886, Plap1, Slrr1c
Chromosome Location
Chr. 13: 49544443 - 49567565 on Build GRCm38
UniGene ID
Mm.383216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172481.1 NP_001165952.1 4 832
NM_025711.3 NP_079987.2 4 849
GenBank mRNA AF316825.1 - 3 569
AK014504.1 - 4 805
AK085920.1 - 4 804
BC034888.1 - 4 751 1365428
BC145904.1 - 4 747 40131207
BC145930.1 - 4 730 40131233

View Full Product Details