Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025793.3 | NP_080069.2 | 7 | 828 | |
GenBank mRNA | AK002352.1 | - | 7 | 789 | |
AK013170.1 | - | 7 | 778 | ||
AK146101.1 | - | 7 | 829 | ||
AK151975.1 | - | 7 | 790 | ||
AK168603.1 | - | 7 | 781 | ||
AK170602.1 | - | 7 | 775 | ||
BC004595.1 | - | 7 | 772 | 3584839 |