Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168320.1 | NP_001161792.1 | 3 | 355 | |
NM_006675.4 | NP_006666.1 | 4 | 422 | ||
XM_011520912.2 | XP_011519214.1 | 5 | 539 | ||
XM_011520913.2 | XP_011519215.1 | 2 | 281 | ||
GenBank mRNA | AF089749.1 | - | 4 | 365 | |
AK027366.1 | - | 4 | 398 | ||
AK223179.1 | - | 4 | 394 | ||
BC071881.1 | - | 3 | 337 | 6256950 | |
CR542109.1 | - | 2 | 239 |