Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025827.3 | NP_080103.1 | 1 | 309 | |
GenBank mRNA | AK004843.1 | - | 1 | 223 | |
AK050231.1 | - | 1 | 218 | ||
AK088348.1 | - | 1 | 310 | ||
AK151103.1 | - | 1 | 255 | ||
AK151406.1 | - | 1 | 249 | ||
AK152964.1 | - | 1 | 249 | ||
AK168628.1 | - | 1 | 259 | ||
AK168791.1 | - | 1 | 255 | ||
AK168956.1 | - | 1 | 255 | ||
AK169656.1 | - | 223 | |||
AK200906.1 | - | 1 | 21 | ||
BC049090.1 | - | 1 | 245 | 6401542 |