Gene Transcripts

Gene Symbol
Vps28
Entrez Gene ID
66914
Gene Name
vacuolar protein sorting 28 (yeast)
Gene Aliases
1110014J03Rik, CIIA, D730005C08Rik
Chromosome Location
Chr. 15: 76622088 - 76626027 on Build GRCm38
UniGene ID
Mm.30028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF373710.1 - 4 204
AK003572.2 - 4 234
AK003699.1 - 4 158
AK013852.1 - 4 155
AK085669.1 - 4 283
BC013535.1 - 4 167 4239189
BY083019.1 - 4 216

View Full Product Details