Gene Transcripts

Gene Symbol
Tmem88
Entrez Gene ID
67020
Gene Name
transmembrane protein 88
Gene Aliases
2600017H02Rik
Chromosome Location
Chr. 11: 69396516 - 69398234 on Build GRCm38
UniGene ID
Mm.390406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025915.4 NP_080191.3 2 437
GenBank mRNA AK010789.1 - 2 444
AK011240.1 - 2 438
AK045149.1 - 2 438
AK161789.1 - 2 438
BC100316.1 - 2 426
BC120912.2 - 2 438 40091214

View Full Product Details