Gene Transcripts

Gene Symbol
Gp2
Entrez Gene ID
67133
Gene Name
glycoprotein 2 (zymogen granule membrane)
Gene Aliases
2310037I18Rik, AV060639
Chromosome Location
Chr. 7: 119442542 - 119459272 on Build GRCm38
UniGene ID
Mm.46403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025989.3 NP_080265.2 3 369
GenBank mRNA AK009323.1 - 3 370
AK009661.1 - 3 370
AK009843.1 - 3 323

View Full Product Details