Gene Transcripts

Gene Symbol
Uqcr10
Entrez Gene ID
66152
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Gene Aliases
1110020P15Rik, AA960494, Ucrc
Chromosome Location
Chr. 11: 4701968 - 4704362 on Build GRCm38
UniGene ID
Mm.269736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197979.2 NP_932096.1 2 309
GenBank mRNA AK002227.1 - 2 299
AK003881.1 - 2 306
AK012489.1 - 2 302
AK168491.1 - 2 283
AW985484.1 - 2 277
BC024518.1 - 2 293 4984172
BC087874.1 - 2 299 6545811
BC100591.1 - 2 299 30119194
BX522336.1 - 3 359
CB599515.1 - 2 312

View Full Product Details