Gene Transcripts

Gene Symbol
Cngb3
Entrez Gene ID
30952
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
CCNC2, CNG6, Cngbeta2
Chromosome Location
Chr. 4: 19280850 - 19510623 on Build GRCm38
UniGene ID
Mm.445778
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013927.2 NP_038955.1 4 492
GenBank mRNA AJ243572.1 - 4 492

View Full Product Details