Gene Transcripts

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
Chromosome Location
Chr. 11: 60971641 - 61020377 on Build GRCh38
UniGene ID
Hs.744366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254750.1 NP_001241679.1 5 1207
NM_001254751.1 NP_001241680.1 5 1207
NM_006725.4 NP_006716.3 5 1207
NR_045638.1 5 1207
XM_006718738.1 XP_006718801.1 5 1207
XM_006718739.1 XP_006718802.1 5 1207
XM_006718740.1 XP_006718803.1 5 1207
XM_006718741.1 XP_006718804.1 5 1207
XM_011545360.1 XP_011543662.1 5 1207
GenBank mRNA AK128419.1 - 6 1111
AK292704.1 - 5 1083
AK312749.1 - 5 1168
BC033755.1 - 5 1123 5213263
BC078669.1 - 5 1146 6302947
U66142.1 - 5 982
U66145.1 - 5 982
U66146.1 - 5 982
X60992.1 - 5 1102

View Full Product Details