Gene Transcripts

Gene Symbol
B3gnt2
Entrez Gene ID
53625
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Gene Aliases
AA408337, AA409404, AW260308, B3Galt6, B3gnt1, BGnT-1, BGnT-2, BGnT1, BGnT2, Beta3gnt
Chromosome Location
Chr. 11: 22834739 - 22860336 on Build GRCm38
UniGene ID
Mm.395495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169114.1 NP_001162585.1 2 1249
NM_016888.5 NP_058584.3 2 1372
GenBank mRNA AF092050.2 - 2 1150
AK160088.1 - 2 1341
AK194572.1 - 1 150
AY043479.1 - 2 1330
BC009075.1 - 2 1249 2654354

View Full Product Details