Gene Transcripts

Gene Symbol
Mkrn1
Entrez Gene ID
54484
Gene Name
makorin, ring finger protein, 1
Gene Aliases
RFP
Chromosome Location
Chr. 6: 39397821 - 39420369 on Build GRCm38
UniGene ID
Mm.270484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018810.2 NP_061280.2 3 539
GenBank mRNA AB026986.1 - 3 644
AF192785.1 - 3 564
AK005137.1 - 3 373
AK029500.1 - 3 579
AK077060.1 - 3 540
AK078962.1 - 3 469
AK186489.1 - 1 147
AK207305.1 - 1 147
AK210725.1 - 1 147
BC003329.1 - 3 528 3158135

Gene Symbol
Mkrn1-ps1
Entrez Gene ID
353327
Gene Name
makorin, ring finger protein 1, pseudogene 1
Gene Aliases
Chromosome Location
Chr. 5: 88837799 - 88839399 on Build GRCm38
UniGene ID
Mm.423672
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF494488.1 - LIT1345
Makorin1-p1
1 352

View Full Product Details