Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018831.4 | NP_061301.3 | 3 | 1801 | |
GenBank mRNA | AF241240.1 | - | 2 | 1272 | |
AK220214.1 | - | 2 | 1029 | ||
BC031858.1 | - | 1 | 692 | ||
BC072667.1 | - | 2 | 1217 | 30629417 | |
BC076617.1 | - | 3 | 1801 |