Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018888.4 | NP_061376.2 | 10 | 2536 | |
GenBank mRNA | AF253516.1 | - | 10 | 2284 | |
AK010376.1 | - | 10 | 2277 | ||
AK139488.1 | - | 10 | 2536 | ||
AK169413.1 | - | 10 | 2273 | ||
AK186801.1 | - | 1 | 294 | ||
AK210478.1 | - | 1 | 55 | ||
AK214249.1 | - | 1 | 92 | ||
AK218492.1 | - | 1 | 55 | ||
AK219516.1 | - | 1 | 55 | ||
BC050884.1 | - | 10 | 2489 | 6808756 | |
BC057570.1 | - | 10 | 2489 | 6808756 |