Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019483.5 | NP_062356.3 | 5 | 1416 | |
GenBank mRNA | AF175408.2 | - | 4 | 1094 | |
AK137411.1 | - | 5 | 1417 | ||
AY145520.1 | - | 4 | 1249 | ||
BC119141.1 | - | 4 | 1133 | 8733890 |