Gene Transcripts

Gene Symbol
Ankrd22
Entrez Gene ID
52024
Gene Name
ankyrin repeat domain 22
Gene Aliases
5430429D21Rik, D19Ertd675e
Chromosome Location
Chr. 19: 34122549 - 34166041 on Build GRCm38
UniGene ID
Mm.183030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024204.6 NP_077166.4 6 963
GenBank mRNA AK012869.1 - 6 977
AK017360.1 - 6 964
AK017408.1 - 6 848
AK037242.1 - 6 846
BC003460.1 - 6 824 2648395

View Full Product Details